Virus Recombinant SARS-CoV-2 Nucleocapsid (N) Protein – C-terminal domain (CTD) (from Insect cell), His Tag
Supplier: ReVacc Scientific
M47G62-100UG M47G62-500UG
76729-560EA 606.58 CAD
76729-560 76729-614
Virus Recombinant SARS-CoV-2 Nucleocapsid (N) Protein – C-terminal domain (CTD) (from Insect cell), His Tag
Proteins and Peptides
A recombinant form of nucleocapsid (N) protein C-terminal domain (CTD) (AA 255-364) from SARS-CoV-2 virus (Wuhan Strain, GenBank: NC_045512.2) was produced by insect cells, followed by purification. This product contains N-terminal Myc tag and C-terminal 6 hexa-histidine tag.
We evaluated the binding of this protein to mAb clone RV20 (NTD-specific, Cat# W89S59), two tag-specific antibodies (anti-His or Myc) and one convalescent serum in ELISAs.