Anti-DYKDDDDK Tag Rabbit Monoclonal Antibody [clone: 114F12C8]

Supplier: Genscript
GenScript®
A01868-40
76198-912EA 348.47 CAD
76198-912
Anti-DYKDDDDK Tag Rabbit Monoclonal Antibody [clone: 114F12C8]
Antibodies
GenScript MonoRab™ DYKDDDDK tag antibody, mAb, rabbit specifically recognizes DYKDDDDK tags placed at C-terminal, N-terminal and internal regions of DYKDDDDK fusion proteins.

  • Comprehensive pool
  • against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection

GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.

Type: Tag
Antigen: DYKDDDDK Tag
Clonality: Monoclonal
Clone: 114F12C8
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR