Anti-SARS-CoV-2 RBD Mouse Monoclonal Antibody [clone: 6D11F2]

Supplier: Genscript
A02055-1 A02055-100
76510-146EA 5052.37 CAD
76510-146 76510-148
Anti-SARS-CoV-2 RBD Mouse Monoclonal Antibody [clone: 6D11F2]
Antibodies
The product is specific for SARS-CoV-2 Spike Protein S1 subunit and its RBD domain.

  • COVID-19 antibodies
  • Validated for ELISA, WB, FACS, and SARS-CoV-2 virus neutralization assay
  • Control antibodies and various antibody pairs available for ELISA-based detection
  • Neutralizing Antibody Standard to calibrate quantitative assay of SARS-CoV-2 neutralizing antibodies in samples

Genscript offers site-specific Neutralizing antibodies, neutralizing antibody standards, and control antibodies.

Type: Primary
Antigen: SARS-CoV-2 RBD
Clonality: Monoclonal
Clone: 6D11F2
More Product Information
Order Now

SPECIFICATIONS

Antigen symbol SARS-CoV-2 RBD
Antigen name SARS-CoV-2 RBD
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 6D11F2
Reactivity
Host Mouse
Isotype IgG1 kappa
ELISA Yes
Flow cytometry Yes
Neutralisation Yes
Immunogen Recombinant SARS-CoV-2 Spike protein fragment
Form Liquid
Purification Protein A affinity column
Concentration 1 mg/ml
Cross adsorption No
Storage buffer PBS, pH 7.4
Storage temperature Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed Avoid repeated freezing and thawing cycles
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR