Stand and Holder for HandyStep® touch Pipette, BrandTech
Supplier: Brandtech
705230 705235
76338-260EA 316.02 CAD
76338-260 76338-264
Stand and Holder for HandyStep® touch Pipette, BrandTech
Pipette Stands
This stand for the HandyStep® touch pipette securely holds the device, even with a PD-Tip II inserted, and saves space on the laboratory bench. It can be used for both the HandyStep® touch and the HandyStep® touch S.
Holder can be attached to a shelf or to a wall (adhesive back).