Hanna Instruments Alkalinity Test Kit
Supplier: HANNA INSTRUMENTS CANADA CA
Hanna Instruments
HI3811 HI3811 HI3811
CA470349-860EA 71.99 CAD
CA470349-860 470006-742 470349-860
Hanna Instruments Alkalinity Test Kit
Environmental Test Kits
This titration-based chemical test kit determines the alkalinity concentration in samples within a 0 to 100 mg/L (ppm) CaCO₃ or 0 to 300 mg/L CaCO₃ range.
  • Supplied with all of the necessary reagents and equipment to perform the analysis
All required materials are included with the test kit, such as the sample beakers, plastic syringe, phenolphthalein indicator, and bromophenol blue indicator.
Method: Phenolphthalein/bromphenol blue.
Ordering information: The test kit contains enough reagents for perform approximately 110 tests. Replacement reagents sold separately.