Refrigerator, Full Stainless Door with Lock

Supplier: Nordon/Hoshizaki Llc.
R1A-FS R2A-FS
76808-998EA 15733.36 CAD
76808-998 76809-000
Refrigerator, Full Stainless Door with Lock
Refrigerators
The Steelheart Series upright single and two section refrigerator with full stainless doors is an attractive workhorse for any business. The dependable ¹/₄ and ¹/₂ HP motor that powers the compressor in the top of this unit provides the cabinet with a continuous stream of cold air through its unique ‘ducted’ air distribution system, which ensures a steady constant temperature.

  • Lockable Full Stainless Door
  • The R1A-FS Includes 3 epoxy coated wire
  • The R2A-FS Includes 6 epoxy coated wire
  • Epoxy electrocoated (E-Coat) evaporator coil to help fight corrosion for extended life
  • Interior LED light bar is mounted horizontally on the ceiling
  • Stainless steel exterior (front, sides, top) and a stainless steel interior

The durable Stainless steel exterior (front, sides, top) and a stainless steel interior resists the random damage normal in a commercial kitchen or business.

The epoxy coated wire shelves adjustable in 1″ increments on stainless steel supports, and the (4) 4″ stem casters (2 with brakes) are standard. It features field reversible, spring assisted self closing door with stay open feature (no kit required).

Electrical Connection: 5 - 15P NEMA, 115 V/60/1, 10 ft. cord and plug
Voltage Range: 104 - 126 V
Control Setpoint Range: 31 to 52 °F
Refrigerant: R290

Delivery information: Five years compressor warranty, three years parts and labor warranty.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR