Sanitary Cubelet Ice Machine and Water Dispensers
Supplier: Nordon/Hoshizaki Llc.
DCM-300BAH DCM-300BAH-OS
76808-988EA 21340.19 CAD
76808-988 76808-990
Sanitary Cubelet Ice Machine and Water Dispensers
Ice Makers
The Hoshizaki sanitary cubelet ice machine and water dispensers feature either LED touch buttons (76808-988) or Opti-Serve touchless operation (76808-990). Both models feature a durable stainless steel exterior, advanced CleanCycle24™ design, and stainless steel auger with greaseless bearing.
  • Up to 321 lbs. of ice production per 24 hours
  • Up to 40 lbs. of built-in storage capacity (1.3 ft.³)
  • 2 second flush cycle every hour
  • Refrigerant: R-404A refrigerant
  • Power: 10.25 A, 6.84 kW
  • Electrical: 115 V, 60 Hz, 1 phase
  • Dispense ice and water
The units canMore Product Information