ABS® Premium Laboratory Refrigerators
Supplier: Horizon Scientific
ABT-HC-PL-23 ABT-HC-PL-49 ABT-HC-SPL-23 ABT-HC-SPL-49
76808-814EA 16693.98 CAD
76808-814 76808-816 76808-822 76808-824
ABS® Premium Laboratory Refrigerators
Refrigerators
ABS Premium Laboratory Refrigerators deliver next level cooling to laboratory environments.
  • Energy saving VSC compressors
  • Microprocessor temperature controllers
  • Ultra-quiet operation
  • Fully array of alarms
  • Controller battery backup
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
Engineered with variable speed compressors (VSCs), these units feature ultra-quiet operation and significant energy savings. VSCs optimize energy consumption by self-adjusting to cooling demands. These compressors also deliver enhanced system performance and provide a longer lifespan than other compressor variations.
Enjoy the uniformity and speed of microprocessor temperature control and a full array of alarms to safeguard your products. Unit controllers also come with a battery backup to keep data safe. Upgrade your laboratory environment with these premium refrigerators and welcome energy savings, noise reduction, smooth operation, and improved system performance in the work environment.
Certifications: ETL Listed.
*Select models ENERGY STAR certified: 76808-814, 76808-816, 76808-822, and 76808-824