Rat NFkB p105/p50 ELISA Kit

Supplier: Antibodies.com LLC
A79563-96
76729-870EA 984.58 CAD
76729-870
Rat NFkB p105/p50 ELISA Kit
Assays ELISAs
Rat NFkB p105/p50 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat NFkB p105/p50 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 0.156 to 10 ng/ml
  • Sensitivity: 0.094 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat NFkB p105/p50 ELISA kit (A79563) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat NFkB p105/p50 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for NFkB p105/p50 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the NFkB p105/p50 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-NFkB p105/p50 Antibody, which binds the captured NFkB p105/p50 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of NFkB p105/p50 captured in each well. The concentration of NFkB p105/p50 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR