Eisco® Zinc-Casted Slotted Mass Hanger, 10 to 100 g
Supplier: Wards
PHZNW050H PHZNW100H PHZNW010H
470345-036EA 10.99 CAD
470345-036 470345-076 470345-496
Eisco® Zinc-Casted Slotted Mass Hanger, 10 to 100 g
Weights Calibration Weights
Zinc-casted slotted mass hanger for use in physics experiments and demonstrations.
  • 10, 50, or 100 g hangers designed for use with slotted mass sets
  • Die-cast in zinc and powder-coated to ensure durability and long life
  • A great financially-friendly alternative to expensive brass hangers
  • Hanger has a ±2% accuracy
  • Serves as a foundation for any set of 10 g slotted masses
These zinc-casted hangers are powder-coated for durability and serve as the foundation for any set of 10 g slotted masses. With a ±2% accuracy, they are an excellent alternate for expensive brass weights.