VWR® Adhesive Sealing Films for PCR and Robotics
Supplier: VWR International
76539-430 76539-432
76539-430PK 111.11 CAD
76539-430 76539-432
VWR® Adhesive Sealing Films for PCR and Robotics
Seals Microplate Seals
Features an extra strong adhesive to minimize evaporation, yet removes easily without leaving a residue on the plate.
  • Certified DNase- and RNase-free
  • Not recommended for qPCR
Standard cut features chamfered corner to eliminate plate overhang for robotics applications and tight storage conditions. Standard cut is ideal use with raised rim and non-skirted PCR plates.
Large cut design provides great plate coverage and easier positioning of the film on the plate. Large cut is recommended for use with flat-top, semi-skirted and full-skirted PCR plates as well as deep-well plates.