VWR® General Purpose Sealing Films
Supplier: VWR International
76539-434 76539-436
76539-434PK 142.16 CAD
76539-434 76539-436
VWR® General Purpose Sealing Films
Seals Microplate Seals
These general purpose seals are ideal for use with polypropylene plates. They minimize evaporation and adhesive plate residue.
  • Certified DNase- and RNase-free
  • Provides a very secure seal to minimize evaporation
  • Can be used as a temporary seal while awaiting further processing
Provides superior barrier properties to minimize evaporation and prevent well to well contamination. The less rigid PP film reduces aerosols upon removal and has a specially formulated adhesive to minimize plate residue.