Kitronik Inventor's Kits
Supplier: NEWARK ELECTRONICS
5603 5603-20
470338-530EA 75.45 CAD
470338-530 470338-532
Kitronik Inventor's Kits
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Build your own electronics project with this breakout board for micro:bit.
  • Quick start: only a screwdriver is required
  • Kit contains breadboard and plenty of electronics to build exciting projects
  • Detailed tutorial book with 10 starter projects included
The Kitronik Inventor's kit for the BBC micro:bit is a great way to get started with programming and hardware interaction with the BBC micro:bit. This Inventor's Kit contains everything you need to complete 10 experiments including using LEDs, motors, LDRs and capacitors.
To get you off to a flying start, we have included an easy-to-follow tutorial book which guides you through everything you will need to know about programming the BBC micro:bit. You don't need any experience with programming as the tutorial book will guide you every step of the way.
The BBC micro:bit has a selection of pins that are located on the bottom edge of its PCB. By using our specially designed Edge Connector Board in conjunction with the breadboard, it is easy to use these pins to connect additional components to the BBC micro:bit.
Ordering information: Required but not included: micro:bit controller.