3D Printing Filament Care Accessories
Supplier: INKSMITH LIMITED SE
3DP-04-0403 3DP-04-0404
470348-380EA 71.09 CAD
470348-380 470348-358
3D Printing Filament Care Accessories
Printer Ribbons
Get the most out of your filament with these accessories.
  • These products are compatible with most 0.5/1 kg, 1.75/2.85 mm filaments on the market
  • The eBox features different settings for different filaments and a timer for auto-shutoff
  • The eVacuum kit is BPA-FREE and made with food-grade materials
The eSun eBox Lite filament dryer regulates the temperature and humidity of your filament spools while in use. Over time, filaments exposed to air can absorb moisture, which leads to degradation and cracking. Once a spool of filament is opened, it must be used quickly to achieve top-quality prints. The eSun eBox filament dryer preserves filament quality and extends the life of filaments once they are in use.
The eSun eVacuum kit is designed for storing filament spools that have been opened but are not currently in use. This kit is perfect for classroom environments, where extended breaks from school might leave a 3D printer idle for weeks or months. Pack your partially-used spools of filament in these vacuum bags to preserve them until they're needed again.
Delivery information: eBox includes (1) dryer storage/dispensing box, (1) feeder tube, (1) power cord. eVacuum kit includes (1) air pump, (10) vacuum bags, (15) humidity indicator cards, (15) desiccant packs, and (2) sealing clips.