Pith Ball Electroscopes
Supplier: American Scientific
3134-01 3134-02
470336-558EA 8.9 CAD
470336-558 470336-560
Pith Ball Electroscopes
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Simple, timeless demonstration of electrostatics.
  • Comes with a teacher’s guide
  • Additional pith balls available in packs of 10
Use this item for demonstrating the attraction and repulsion of electric charges. It consists of an insulated brass pillar and hook on a base. It is useful for interesting and engaging demonstrations.
Delivery information: Supplied with two colored pith balls with strings attached.