MyPIPETMAN® Select Pipettors
Supplier: GILSON, INC.
FP10011S FP10012S FP10013S FP10014S FP10015S FP10016S FP10001S FP10002S FP10003S FP10004S FP10005S FP10006S FP10007S FP10008S FP10009S FP10010S
76716-740EA 2150.1 CAD
76716-740 76716-742 76716-744 76716-746 76716-748 76716-750 76716-720 76716-722 76716-724 76716-726 76716-728 76716-730 76716-732 76716-734 76716-736 76716-738
MyPIPETMAN® Select Pipettors
Pipettes Micropipettes
Ergonomic, safe and reliable, MyPIPETMAN® Select offers volume setting choice and optional handle personalization for optimal use in various protocols.
  • Variable volume single channel models covering a volume range from 0.2 μl to 10 ml
  • Variable volume multichannel models covering a volume range from 0.5 μl to 300 μl (8 and 12-channel models)
  • Free volume setting position for quick and effortless volume adjustment
  • Light ratchet volume setting for precise volume adjustment
  • Volume locked position to secure pipetting
Set and secure pipetting volumes to meet specific needs and requirements. Simply adjust the volume by turning the push button of the pipette with the unique, patented Trilock™ volume-locking system. Lightweight and well-balanced, MyPIPETMAN® Select has an elegant and ergonomic design with a textured effect for a better grip. Without compromising on quality and performance, the built-in ejection system with an integrated lever provides reduced tip ejection forces compared to direct ejection systems. This patented ejection system reinforces the pipette’s ergonomics for convenient use throughout the day.
The MyPIPETMAN® Select offers optional handle printing on demand. Identify the pipette to avoid any risk of contamination by dedicating it to a specific liquid or workflow step and improve the lab's lean management process.