ActivArmr® Electrical Insulating Gloves, Class 00
Supplier: Ansell Healthcare
RIG0011R085 RIG0011R090 RIG0011R095 RIG0011R100 RIG0011R105 RIG0011R110 RIG0011R120
76683-118EA 124.41 CAD
76683-118 76683-126 76683-140 76683-134 76683-116 76683-136 76683-130
ActivArmr® Electrical Insulating Gloves, Class 00
Gloves
Electrical insulating gloves are made from natural rubber latex using a proprietary environmentally friendly dipping process for ultimate flexibility and dexterity.
  • Natural rubber latex for ultimate durability and flexibility
  • Ergonomically-designed hand-at-rest shape for less hand fatigue
  • Generous flared cuff allows room for clothing and improves ventilation
  • Case hardened finish provides smoother surface for easy donning and doffing performance and protection
  • Product meets all applicable NFPA, OSHA and CSA standards
  • ISO 14001 certified manufacturing facility
Electrical insulating gloves feature an ergonomic hand-at-rest shape and non-splayed fingers to reduce hand fatigue. The generous flared cuff allows room for clothing, and a smooth finish allows for easy donning and doffing.
Electrical glove Class 00 offers arc flash protection Class 1 in accordance with EN 61482-1-2. Certified for resistance to acid (Category A), Ozone (Category Z), and very low temperatures (Category C). Applications with risk of touch voltage AC maximum 500 V (DC maximum 750).
Certifications: CE marked, PPE category ELECTRICAL, Category III, EN 388:2016, EN 420:2003 + A1:2009, EN 60903:2003, Arc Rating of APC 1, ASTM D120, REACH compliant.
Packaging: Gloves are packaged 1 pair per bag, 10 pairs per carton.