Eisco Sterile Polystyrene Petri Dishes with Lids
Supplier: Wards
CH0374BPK10 CH0374APK10
470344-384EA 11.55 CAD
470344-384 470345-044
Eisco Sterile Polystyrene Petri Dishes with Lids
Culture Dishes Petri Dishes
Individual packed and sterile disposable Petri dishes with lid made of transparent polystyrene with triple vents.
  • Made of high quality transparent polystyrene
  • Transparent for clear examination of contents
  • Individually packed in polyethylene bags and sterilized by radiation
  • Non-toxic, multifunctional and ready to use
  • Triple vented to allow for airflow and discourage condensation build-up
  • Ideal for applications requiring better gaseous exchange or evaporation
Triple vented lid to allow for airflow and discourage condensation build-up. Ideal for applications requiring better gaseous exchange or evaporation. Petri dish is packed individually in polyethylene bags and sterilized by radiation. Excellent for general laboratory applications, including cell culturing, plant germination, precipitate drying and solvent evaporation, home or classroom science experiments, crafting, small storage solutions and more. Pack of 10 plates.