Eisco Vacuum Desiccators with PTFE Plug and Stopcock
Supplier: Wards
CH0366C CH0366B CH0366A
470344-374EA 342.4 CAD
470344-374 470344-498 470346-118
Eisco Vacuum Desiccators with PTFE Plug and Stopcock
Desiccators Desiccators, Plastic
Polypropylene and polycarbonate vacuum desiccator with flat dome, with PTFE plug and stopcock, silicone rubber O-ring on the flange.
  • Holds a full vacuum (740 mm Hg 15 cm) at room temperature for 24 hours - will remain airtight even if not under vacuum
  • Clear PC lid on a chemical-resistant white PE base
  • Internal groove on the flange holds a silicone rubber O-ring
  • Fitted with a polypropylene stopcock with a self-lubricating PTFE plug for 3-way action: vacuum creation, shut off, and releasing
  • 'Flat dome' features clear polycarbonate top with large ribbed knob
More Product Information