Eisco Graduated Micro Centrifuge Tubes
Supplier: Wards
CH0284A CH0284B
470344-523PK 105.7 CAD
470344-523 470345-987
Eisco Graduated Micro Centrifuge Tubes
Tubes Microcentrifuge Tubes
Pack of 100 graduated, leak-proof micro centrifuge tubes made of polypropylene with raised graduations and snap cap lids.
  • Made of high quality polypropylene
  • Raised graduations
  • Features built in plugs which snap to close the top for a leak proof seal
  • Can withstand high forces of an ultra centrifuge and can be used for transmitting samples for micro analysis
  • Each tube is 1.25" long and 0.3" wide
Pack of 100 Micro Centrifuge Tubes made of high quality polypropylene with raised graduations. Features built in plugs which snap to close the top for a leak proof seal.