Corepoint® Scientific White Diamond Flammable Material Storage Touchscreen Manual Defrost Freezers
Supplier: Horizon Scientific
FF141WWW/0MHCTS FF171WWW/0MHCTS FF201WWW/0MHCTS
76717-360EA 16400.24 CAD
76717-360 76717-364 76717-366
Corepoint® Scientific White Diamond Flammable Material Storage Touchscreen Manual Defrost Freezers
Freezers Flammable Material Freezers
Safeguard volatile substances and upgrade your performance with these specially-designed flammable storage freezers.
  • Laboratory use
  • Microprocessor temperature controller
  • Temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These White Diamond Series Flammable Storage Freezers meet the requirements of NFPA 45 and NFPA 30 for the cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsicallyMore Product Information