Human Heat Shock 70 kDa Protein 4L(HSPA4L) ELISA Kit
Supplier: AFG Bioscience
EK712681
76704-082EA
601.32
CAD
76704-082
Human Heat Shock 70 kDa Protein 4L(HSPA4L) ELISA Kit
Assays
ELISAs
- Detection wavelength: 450 nm
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...