Thermo Scientific™ Matrix™ 2D Barcoded Screw Top Storage Tubes, 12 ml
Supplier: Thermo Fisher Scientific
3770 3775 3776 3778
76642-906CS 645.48 CAD
76642-906 76642-820 76642-938 76642-684
Thermo Scientific™ Matrix™ 2D Barcoded Screw Top Storage Tubes, 12 ml
Vials
Enable safe, secure tracking of your precious samples with Thermo Scientific™ Matrix™ 2D barcoded 12 ml screw top storage tubes, the perfect solution for large-volume storage, including clinical samples, powders, tissue and plant materials. 
  • Laser-etched 2D barcode on the bottom of each tube allows you to scan and decode tubes at once without removing them from the rack, it also allows data to be associated with each tube
  • Thermo Scientific™ VisionMate™ Lite or VisionMate wireless can be used to read single tubes, or the VisionMate HighSpeed can be used to decode an entire rack of tubes instantly
  • Tube can be scanned into a database or tracking system or integrate via ActiveX technology
Superior storage format:
Supplied in microplate footprint rack to store more samples in less freezer space
Compatibile with Matrix™ Electronic EXP Pipettes
Locking rack lid for secure storage of samples
Rack base and tube design to prevent the tube from spinning in the rack when decapping
Solid, one-piece construction that enables storage down to cryogenic temperatures
Stringent quality control:
Every 2D barcode is scanned for readability and checked against a full database of all previous 2D barcodes to ensure there are zero duplicates across the entire Thermo Scientific Matrix™ line of storage tubes
Each tube is 100% leak tested to ensure tube integrity and sample security
Built-in error correction ensures that barcodes will never be misread
Certifications: Products are manufactured in ISO 7 class clean room environment according to ISO 14644-1:2015. Sterile products are sterilized by gamma irradiation according to ISO 11137; 10⁻³ Sterility Assurance Level (SAL). The vials and closures were tested and met the requirements of USP <88> Class VI plastics.