Thermo Scientific™ Matrix™ 2D Barcoded Low Volume Screw Top Storage Tubes
Supplier: Thermo Fisher Scientific
3747 3748 3748-BR
76642-618CS 753.46 CAD
76642-618 76642-676 76642-674
Thermo Scientific™ Matrix™ 2D Barcoded Low Volume Screw Top Storage Tubes
Vials
Maximize storage capacity with the space-saving Thermo Scientific™ Matrix™ 2D barcoded low volume screw top storage tubes, now available in a barcoded rack.
  • Total height of less than 23 mm — manual or automated storage capacity can be as much as doubled compared to higher volume tube options
  • Maximizing storage capacity allows improved utility of laboratory space and energy consumption
  • 200 μl 2D internally-threaded universal cap features single-piece construction with molded-in-gasket that cannot be over-tightened manually or with automated capping tools
  • Thermoplastic elastomer gasket material is compatible for storage at room temperature down to the vapor phase of liquid nitrogen
These 200 μl 2D internally-threaded tubes maximize capacity in manual or automated storage systems. They can be used for storage in virtually any laboratory condition, from room temperature through the vapor phase of liquid nitrogen. Permanent 2D barcodes enable safe, Secure tracking of valuable small-volume samples. Compatible with the complete line of Thermo Scientific 2D barcode readers, sample data can be integrated into existing database or tracking systems.
Secure tracking:
Permanently-bonded, unique 2D barcode is laser-etched onto the base of every Thermo Scientific Matrix™ 2D barcoded screw top storage tubes for secure identification and sample tracking
A Thermo Scientific 2D barcode reader instantly decodes each tube's 2D barcode scanning the tube into any application or database, or integrates via several export options
New barcoded rack allows tracking of the rack in addition to tubes
Laser-etched barcodes and human readable text on three sizes of rack, plus 2D code on rack bottom
Once sample data is stored, just scan the tube to access sample information and identify the contents
Stringent quality control:
Every 2D barcoded storage tube is scanned to ensure readability
All barcodes are checked against our database of previously assigned codes to prevent duplicates
Every tube is leak tested to ensure sample integrity and security
Certifications: Products are manufactured in ISO 7 class clean room environment according to ISO 14644-1:2015. Sterile products are sterilized by gamma irradiation according to ISO 11137; 10⁻³ Sterility Assurance Level (SAL). The vials and closures were tested and met the requirements of USP <88> Class VI plastics.