Thermo Scientific™ Sample Storage Starter Packs
Supplier: Thermo Fisher Scientific
HHD-HSX-WP1D-2 HHD-HSX-WP1D-505 HHD-ST-WP1D-2 HHD-WP1D-2 HHD-HSX-WP1D-510
76642-822EA 17991.46 CAD
76642-822 76642-702 76642-644 76642-914 76894-076
Thermo Scientific™ Sample Storage Starter Packs
Vials
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs.
  • Eliminate the need to reformat non-2D cryotube samples
  • Secure sample tracking with 2D, 1D, and human readable codes
  • Scan 96 tubes in as little as 1 second with advanced and premium starter packs
  • Help ensure quality with ISO-7 class cleanroom– manufactured tubes with sterility assurance
Choose from five different starter pack options.