GenScript® Magnetic Beads for Protein and Antibody Purification
Supplier: Genscript
L00695-4 L00895-10 L00895-2 L00936
76510-452EA 1032.37 CAD
76510-452 76683-230 76683-228 76683-232
GenScript® Magnetic Beads for Protein and Antibody Purification
Beads Magnetic Beads
To enable fast and efficient purification of antibody and epitope-tagged proteins, GenScript provides super paramagnetic magnetic beads. GenScript magnetic beads are coated with recombinant protein ligands that enable highly specific affinity binding to antibodies and epitope-tagged proteins.
  • Quick, easy, and convenient magnetic separation procedures
  • High binding capacity
  • Reproducible results due to highly consistent bead size
  • Extremely gentle sample handling minimizing protein degradation
The magnetic protein purification method allows for protein purification directly from cell lysates, thus eliminating the need for centrifugation steps and simplifying protein purification. The high binding capacity, specificity of binding, and superfast magnetic response make GenScript magnetic beads ideal for applications requiring fast turnaround time and large number of samples.
The advantage of using magnetic beads is that it eliminates steps like centrifugation or filtration, which are involved in the traditional methods of protein purification. This can significantly save your time and labor.