Thermo Scientific™ Matrix™ Blank and Alphanumeric Storage Tubes
Supplier: Thermo Fisher Scientific
4170 4247 4248 4249 4250 4251 4252-11 4253 4271 4272 4273 4274 4140 4147 4148-11 4150
76642-582CS 201.68 CAD
76642-582 76642-896 76642-540 76642-732 76642-954 76642-942 76642-526 76642-944 76642-856 76642-752 76642-894 76642-910 76642-770 76642-818 76642-908 76642-610
Thermo Scientific™ Matrix™ Blank and Alphanumeric Storage Tubes
Vials
Store valuable samples where 2D tracking is not required with high-density Thermo Scientific™ Matrix Blank and Alphanumeric storage tubes, offered in manual and automation-friendly racks.
  • Matrix storage tubes are manufactured from medical grade virgin polypropylene
  • Tubes are 100% leak tested to ensure sample integrity and security
  • Tubes are validated to be free of DNAse, RNAse, endotoxins and cytotoxins
Visual identification of samples can be made using the alphanumeric code on the bottom of each tube, and the tubes are compatible with the full line of seals and capping equipment. Ideal for scaling up from microplate or DeepWell™ block sample storage.
Superior storage rack design:
Unlike traditional storage tubes or blocks, Thermo Scientific Matrix blank and alphanumeric storage tubes are available in our specially designed, stackable, microplate-footprint Latch Racks of 96 tubes for individual sample, access and removal
The Latch Rack design enables multichannel pipette access and eliminates contamination risk with a lid that never touches the bench top when open
The Latch Rack is also compatible with automated liquid handling, as the lid can be lifted robotically for high-throughput applications
Clear designations:
Matrix alphanumeric storage tubes are clearly marked with high contrast printing on the base of each tube, designating their appropriate well location within a rack, for easy identification
Enhance ease of visual identification with colored SepraSeal capping options or labelling on the side of the tube using the ID Scribe instrument
Capping options:
Several sealing options are available, including the Thermo Scientific Matrix line of CapMats and CapStrips for sealing multiple tubes and the Thermo Scientific Matrix SepraSeal and DuraSeal capping systems for individual tube sealing, access and piercing
Applied with the Thermo Scientific SuperSealer, sealing is secure and consistent every time
Compatible with:
Capping systems, SepraSeals, Duraseals, SuperSealer, ID Scribe
Recommended for:
Individual sample retrieval; Shipping samples; Storage of DNA, compounds, or any other samples
Certifications: Products are manufactured in ISO 7 class clean room environment according to ISO 14644-1:2015.
Sterile products are sterilized by gamma irradiation according to ISO 11137; 10⁻³ Sterility Assurance Level (SAL)
The vials and closures were tested and met the requirements of USP <88> Class VI plastics.