Thermo Scientific™ SuperSealer
Supplier: Thermo Fisher Scientific
4423 4110-11
76642-600EA 60.29 CAD
76642-600 76642-790
Thermo Scientific™ SuperSealer
Film Applicators Sealing Film Applicators
Ensure a proper seal on plates, blocks and storage tube racks with the Thermo Scientific™ SuperSealer. Choose individual sealing options such as Thermo Scientific™ Matrix™ SepraSeal mats for liquid handling and single tube accessibility, or seal entire plates and blocks for storage with Thermo Scientific™ Nunc™ CapMats.
  • Simplifies the sealing of plates, blocks and tube racks of virtually any height, with no adjustments needed, ensuring a secure and consistent seal
  • Once the seal along with the plate, block or tube rack are placed into the drawer, simply close the door to initiate sealing
  • Designed to work with Thermo Scientific™ SepraSeal mats, CapMats and, with the SuperSealer AdhesiveSeal Mat, any pre-cut adhesive tape seal, you can be assured a proper seal regardless of sealing option
Includes: Automatic cap, tape and SepraSeal mat applicator.