Thermo Scientific™ Matrix™ Screw Top Tubes, 0.5 ml
Supplier: Thermo Fisher Scientific
3743 3743AMB 3743BLU 3743GRE 3743PUR 3743RED 3743WHI 3743YEL 3744 3744BLU 3744GRE 3744PUR 3744RED 3744WHI 3744-WP 3744YEL 3745
76642-830CS 705.06 CAD
76642-830 76642-720 76642-570 76642-546 76642-898 76642-646 76642-946 76642-634 76642-802 76642-786 76642-956 76642-566 76642-886 76642-812 76642-624 76642-774 76642-584
Thermo Scientific™ Matrix™ Screw Top Tubes, 0.5 ml
Vials
Store samples securely at low temperatures, including vapor phase Liquid Nitrogen with Thermo Scientific™ Matrix™ 500 µl screw top tubes.
  • Tubes are constructed of virgin class VI medical grade polypropylene
  • ScrewTop caps feature a soft silicone gasket that conforms to mating surfaces to ensure sample preservation
  • Racks are constructed of sturdy acrylonitrile butadiene styrene (ABS) to protect tubes in case of accidental dropping
  • ScrewTop Tubes are compatible with storage down to –196 °C
Used in combination with Thermo Scientific™ VisionMate™ 2DMore Product Information