Thermo Scientific™ Matrix™ Open-Top Tubes in Barcoded Latch Racks
Supplier: Thermo Fisher Scientific
3791-BR 3792-BR 3732-BR 3712-11-BR
76642-760CS 1319.78 CAD
76642-760 76642-560 76642-892 76642-562
Thermo Scientific™ Matrix™ Open-Top Tubes in Barcoded Latch Racks
Vials
Securely store and track samples in Thermo Scientific™ Matrix™ open-top tubes.
  • A unique 2D barcode is laser-etched onto the base of every polypropylene open-top tube to securely identify and track samples
  • 2D barcodes are resistant to 100% DMSO, ideal for compound processing and storage
  • Non-proprietary 2D codes can be scanned with the Thermo Scientific VisionMate family of barcode readers
Laser etching produces a permanent, high-contrast, 2D barcode on each tube, offering maximum reliability and traceability for chemical, biological, and genomic samples. Barcoded latch racks feature a linear barcode, 2D barcode, and human readable rack identification on three sides of the rack. Use in combination with Thermo Scientific™ Matrix™ SepraSeal and DurSeal capping systems and the Thermo Scientific™ VisionMate™ family of barcode readers.
Superior storage format:
ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
Barcoded latch racks include 1D and 2D barcodes, plus human readable text on three sides to provide added same traceability
The latch rack lid can be lifted robotically for 2D tube access with automated liquid handling systems and high throughput applications
Stringent quality control:
Every 2D barcoded storage tube is scanned to guarantee readability.
Each code is checked against a full database of all previously assigned 2D codes to ensure zero duplicates across the entire line of Matrix 2D barcoded storage tubes.
Storage tracking solutions for any laboratory:
Custom 2D coding options are available to meet your laboratory's individual database or tracking requirements
Tubes are available with a range of sealing options including Thermo Scientific SepraSeals and DuraSeals
SepraSeals are available in a range of color options for visual categorization of samples
Pre-Split SepraSeals allow for repeated sample access in automation
Recommended for:
Drug discovery, compound storage, secure sample transport, individual sample retrieval, instant sample identification, Secure tracking of archived samples
Certifications: Products are manufactured in ISO 7 class clean room environment according to ISO 14644-1:2015.
Sterile products are sterilized by gamma irradiation according to ISO 11137; 10⁻³ Sterility Assurance Level (SAL)
The vials and closures were tested and met the requirements of USP <88> Class VI plastics.