Thermo Scientific™ Matrix™ 2D Barcoded Clear Polypropylene Open-Top Storage Tubes with Installed DuraSeal Septum Seal

Supplier: Thermo Fisher Scientific
3736 3737 3729 3801
76642-736CS 1552.11 CAD
76642-736 76642-654 76642-934 76642-876
Thermo Scientific™ Matrix™ 2D Barcoded Clear Polypropylene Open-Top Storage Tubes with Installed DuraSeal Septum Seal
Vials
Store, track and access samples with ease with Thermo ScientificTM MatrixTM 2D Barcoded Clear open-top storage tubes with DuraSeal septum installed.

  • A permanently-bonded, unique 2D barcode is laser-etched onto the base of every Matrix 2D barcoded storage tube to securely identify and track samples
  • Corresponding with a laboratory's throughput needs, Thermo Scientific™ VisionMate™ 2D barcode readers instantly decode each tube's 2D barcode
  • A range of VisionMate 2D barcode readers is available to meet your application and throughput needs
  • Scan the tube into any application or database, or integrate via various methods
  • VisionMate systems are flexible enough to allow 2D barcode data to be integrated into various applications, databases and automated systems
  • Once sample data is stored, user can scan the tube with the database
  • Tracking system includes a 'find' feature to instantly access sample information and identify the contents of each storage tube

The permanently-installed DuraSeal septum allows easy access to tube contents and enables long-term storage at temperatures down to –150 °C. The proprietary barcoding process produces a permanent, high-contrast 2D barcode offering maximum reliability and traceability of compound, biological and genomic samples.

Superior storage rack design:
Matrix 96-format 2D barcoded storage tubes are available in a proprietary specially-designed, stackable, microplate footprint
Latch racks save precious space in your freezer, refrigerator, compound store or on the bench top
The latch rack design enables manual multichannel pipette access to 2D tubes and eliminates contamination risk with lid design that does not contact the benchtop
The latch rack lid can be lifted robotically for 2D tube access with automated liquid handling systems and high throughput applications

Stringent quality control:
Every 2D barcoded storage tube is scanned to guarantee readability
Each code is checked against a full database of all previously assigned 2D codes to ensure zero duplicates across the entire line of Matrix 2D barcoded storage tubes

Storage tracking solutions for any laboratory:
Custom 2D coding options are available to meet your laboratory's individual database or tracking requirements
2D storage tubes are compatible with Thermo Scientific freezer racks that allow optimal utility of storage space

Certifications: Products are manufactured in ISO 7 class clean room environment according to ISO 14644-1:2015.
Sterile products are sterilized by gamma irradiation according to ISO 11137; 10⁻³ Sterility Assurance Level (SAL)
The vials and closures were tested and met the requirements of USP <88> Class VI plastics.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR