Chemical Wash Bottle, Blank Labels
Supplier: Wards
CHWB1006 CHWB1004 CHWB1005
470344-144EA 12.95 CAD
470344-144 470344-978 470345-636
Chemical Wash Bottle, Blank Labels
Bottles Wash Bottles
Chemical wash bottles feature a wide mouth design, and are self-venting. Bottles are made of low-density polyethylene, providing convenience and durability in laboratory applications.
  • Chemical wash bottles with blank labels
  • Blank label provides maximum flexibility, allowing user to write in contents and safety information
  • Wide mouth for easy filling
  • Self venting design to eliminate leaks and dripping
  • Easy to control squeeze for dispensing
Made of low density polyethylene (LDPE) for superior quality and strength. Innovative Flex Ink won't fade, crack, or peel. Blank label provides maximum flexibility, allowing user to label with contents and safety information. Wide mouth for easy refilling. Each bottle features a two-piece construction, consisting of the bottle and leak-proof cap with built-in nozzle. The low density polyethylene plastic is very responsive, with smooth bounce back that won't get stiff over time.