Synthware NMR Tubes, 5 mm
Supplier: Kemtech America
XWE-5MM-7 XWE-5MM-7-50
75876-200CS 24.05 CAD
75876-200 75876-202
Synthware NMR Tubes, 5 mm
Tubes NMR Tubes
Economy grade NMR tubes with caps included.
  • 33 expansion borosilicate glass
  • 7"
  • 100 MHz rating
Designed to tightly fit in any spinner turbine and have zero NMR background.