Synthware Distilling Receivers, Graduated, with Hooks
Supplier: Kemtech America
T230012 T230015
60002-106EA 73.85 CAD
60002-106 60002-108
Synthware Distilling Receivers, Graduated, with Hooks
Distillation Components Distillation Receivers
Centrifuge tube shaped receiver with an upper standard taper joint.
  • Heavy wall construction
  • 33 expansion borosilicate glass
  • Standard ground glass joints work with other brands
Designed for use with D381412 distilling receiver.