Synthware Plug, High Vacuum, Teflon® Protected, Metering
Supplier: Kemtech America
S660008 S660012
60004-052EA 104.07 CAD
60004-052 60004-054
Synthware Plug, High Vacuum, Teflon® Protected, Metering
Stopcocks
Designed to provide an excellent vacuum tight seal without exposing any material other than Teflon® to chemicals.
  • Teflon shaft
  • PTFE covered Viton O-rings
Complete with control knob. Viton® O-rings on PTFE shaft are protected with two precision Teflon® wipers. The plug has a machined-in integral PTFE tip O-ring to make the primary seal.