Synthware Vacuum or Inert Gas Manifolds
Supplier: Kemtech America
M101003 M101004 M101005
60004-418EA 1113.57 CAD
60004-418 60004-420 60004-422
Synthware Vacuum or Inert Gas Manifolds
Manifolds
Vacuum manifold with front-left-right and rear-left 10 mm hose connections.
  • Heavy wall construction
  • 33 expansion borosilicate glass
  • Standard ground glass joints work with other brands
General purpose double bank manifold for use with vacuum and inert gas.
Order Now
SPECIFICATIONS
Environmentally Preferable