Oakton® ORP Electrodes, Cole-Parmer
Supplier: Antyila Scientific
59001-77 35805-15 35805-13 59001-75
76326-440EA 321.75 CAD
76326-440 76326-460 76326-494 76326-502
Oakton® ORP Electrodes, Cole-Parmer
Electrodes Redox Electrodes
Reliable and cost effective probes.
  • Won't break - epoxy electrodes resist impact and are extremely durable
  • Glass electrodes resist aggressive chemicals
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with the sample.
Choose junction and tip style based on your application. All electrodes feature Ag/AgCl reference cells.