United Scientific Aluminum Alloy Tuning Forks
Supplier: United Scientific Supplies
TFK480 TFK341 TFK426 TFK512 TFK640 TFK100 TFK1024 TFK128 TFK2048 TFK256 TFK288 TFK320
470005-910EA 9.8 CAD
470005-910 470349-624 470349-626 470349-674 470349-614 470349-610 470349-648 470349-672 470349-662 470349-642 470349-644 470349-680
United Scientific Aluminum Alloy Tuning Forks
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Long, pure sounding aluminum tuning forks.
  • Aluminum alloy
  • Each fork is individually stamped with the frequency and note
The frequency and scale letter is stamped on each fork. These easy to activate tuning forks are perfect for investigating sound. Do not strike with a hard object, use a tuning fork activator.