VWR® Top Hinge Incubator Shakers, with Optional Refrigeration
Supplier: VWR International
76644-702 76644-704
76644-702EA 12637.16 CAD
76644-702 76644-704
VWR® Top Hinge Incubator Shakers, with Optional Refrigeration
Shakers Shaking Incubators
VWR® Top hinge incubator shakers with optional refrigeration are designed to reliably provide optimal performance to safely shake flasks, tube racks, and microplates in a temperature-controlled environment.
  • Easy to use LED user interface
  • Clear curved tempered glass window for viewing samples without disturbing chamber environment
  • Designed for quiet operation
  • Variety of flask clamps, sticky mats, tube racks and plate holder to accommodate various sample holders
  • Timer, audible and visual alarms
  • 6 programmable segments varying temperature and speed
  • Highly efficient inverted compressor refrigeration system (76644-702 only)
VWR® Top hinge incubator shakers are quiet, easy to operate and convenient to load and are ideal for cell culture, solubility studies, extraction procedures, protein expression and many other applications.
Certifications: CE, UL
Delivery information: Supplied with platform installed, power cord, 2-year warranty.