Eisco Double Convex Glass Lenses
Supplier: Wards
PH0534ECV PH0534OCV PH0534PCV PH0534RCV1 PH0534RCV2 PH0534NCV PH0534HCV PH0534QCV PH0534RCV PH0534ICV PH0534KCV PH0534GCV PH0534JCV PH0534DCV PH0534CCV PH0534BCV PH0534ACV PH0534FCV
470328-066EA 8.7 CAD
470328-066 470328-068 470328-070 470328-072 470328-074 470343-600 470344-090 470344-116 470344-182 470344-354 470344-550 470344-644 470344-646 470345-002 470345-032 470345-212 470345-396 470345-708
Eisco Double Convex Glass Lenses
Educational Materials Physics Educational Materials Optics Learning Activities
Individual spherical glass lenses.
  • Double convex lenses in a variety of diameters and focal lengths
  • Made of high quality, optically worked glass
  • Highly polished surface, ground edges
This optically true lens is excellent for physics classroom demonstrations in optics and light refraction.
Eisco labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control.
Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments. These lenses are designed for educational use only.