Water, GemPure™ Select USP, purified, non sterile

Supplier: GeminiBio

901-006-1X6 901-006-C23 901-006-PD2
CA76683-194CS 145.82 CAD
CA76683-194 CA76683-196 CA76683-198
Water, GemPure™ Select USP, purified, non sterile
Water

GemPure™ Select Purified Water Non-Sterile (USP Grade) is manufactured and tested in accordance with the USP monograph for Purified Water. Purified water is suitable in various non-parenteral applications, non-active medical device manufacture and cleaning applications.


  • Manufactured in an ISO 13485:2016 and 21CFR820 compliant facility
  • Available in 1, 20, 200 L, as well as custom sizes to suit your needs
  • For further manufacturing or research use only
  • Not for direct therapeutic use

Formula: H₂O
MDL Number: MFCD00011332
CAS Number: 7732-18-5
UN: 2201

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR