Water, GemPure™ Select USP for irrigation (WIR)
Supplier: GeminiBio
GeminiBio’s Water for Irrigation (WFIr) Quality Water (USP Grade) is a hypotonic, sterile non-pyrogenic irrigating water produced by our proprietary water purification system. Manufactured and tested in compliance with the USP monograph for bulk packaged water for irrigation, GemPure™ Select Water for Irrigation (WFIr) Quality Water (USP Grade) is ideal for use in the manufacture and testing of medical devices, and applications where Water For Injection is not required.
- Manufactured in an ISO 13485:2016 and 21CFR820 compliant facility
- Available in 500 ml, 1, 20 and 200 L, as well as custom sizes to suit your needs
- For further manufacturing or research use only
- Not for direct therapeutic use
Meets the USP monograph for Water for Irrigation packaged in bulk for commercial use.
Formula:
H₂O |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 UN: 2201 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...