Water, GemPure™ Select USP for injection (WFI)
Supplier: GeminiBio
901-001-5X6
901-001-1X6
901-001-011
901-001-021
901-001-103
901-001-203
CA76683-160CS
175.13
CAD
CA76683-160
CA76683-162
CA76683-164
CA76683-166
CA76683-168
CA76683-170
Water, GemPure™ Select USP for injection (WFI)
Water
GemPure™ Select Water for Injection (WFI) Quality Water (USP Grade) optimizes your upstream and downstream bioprocess workflows and productivity from development to commercial production.
- Manufactured in an ISO 13485:2016 and 21CFR820 compliant facility
- Available in 500 ml, 1, 10, 20, 100 and 200 L, as well as custom sizes to suit your needs
- For further manufacturing or research use only
- Not for direct therapeutic use
Meets the USP monograph for Water for Injection packaged in bulk for commercial use.
Formula:
H₂O |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 UN: 2201 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...