Water, GemPure™ Plus Ph. Eur., JP, USP injection_WFI
Supplier: GeminiBio
Synonyms:
Hydrogen oxide
901-002-4X4
901-002-021
901-002-203
CA76683-172CS
651.66
CAD
CA76683-172
CA76683-174
CA76683-176
Water, GemPure™ Plus Ph. Eur., JP, USP injection_WFI
Water
GemPure™ Plus Water for Injection (WFI) Quality Water (USP/EP/JP Grade) optimizes your upstream and downstream bioprocess workflows and productivity from development to commercial production and meets specifications of monographs for the United States Pharmacopeia (USP), European Pharmacopeia (EP), and Japanese Pharmacopeia (JP).
- Manufactured in an ISO 13485:2016 and 21CFR820 compliant facility
- Available in 4, 20, 200 L, as well as custom sizes to suit your needs
- For further manufacturing or research use only
- Not for direct therapeutic use
Multicompendial, meeting testing specficiations for USP, EP, and JP monographs.
Formula:
H₂O MW: 18.02 g/mol Boiling Pt: 100 Melting Pt: 0 |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 UN: 2201 Merck Index: 13,10098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...