VWR® Vials, Borosilicate Glass with Phenolic Cap and Polyvinyl-Faced Pulp Liner
Supplier: VWR International
76538-970 76538-974 76538-976 76538-978 76538-980 76538-982 76538-984 76538-988 76538-990
76538-970CS 1916.84 CAD
76538-970 76538-974 76538-976 76538-978 76538-980 76538-982 76538-984 76538-988 76538-990
VWR® Vials, Borosilicate Glass with Phenolic Cap and Polyvinyl-Faced Pulp Liner
Vials
Made from borosilicate glass with phenolic cap and polyvinyl-faced pulp liner for general-purpose applications.
  • Short form or capsule style
  • Cap is flush to vial shoulder
  • Black phenolic molded screw cap
  • Polyvinyl-faced pulp liner
  • General-purpose use
The cap on the vial is the same diameter as the body, fitting flush to the shoulder.