VWR® Scintillation Vials with Screw Cap, Borosilicate Glass
Supplier: VWR International
76538-954 76538-956 76538-960 76538-962 76538-964 76538-966
76538-954CS 343.31 CAD
76538-954 76538-956 76538-960 76538-962 76538-964 76538-966
VWR® Scintillation Vials with Screw Cap, Borosilicate Glass
Vials
VWR® scintillation vials are produced from borosilicate 3.3 glass. They are compatible with all scintillation counters.
  • Borosilicate glass construction
  • Fit all standard liquid scintillation counting equipment
  • High resistance against chemicals and thermal shock
  • Low background counts and high ultraviolet transmission
  • Choose attached or unattached caps
VWR® scintillation vials are ideal for use in liquid scintillation counting, beta/gamma counting or for sample storage.
Each vials includes a white screw cap with a top suitable for marking.
76538-962 feature smaller overall dimensions to permit substantial reduction in the quantity of solvent required with resultant savings in solvent cost.
Ordering information: Available with a variety of caps and liners.