sparQ mRNA-Seq Kit
Supplier: Quantabio
95218-024 95218-096
CA76683-154EA 1423.36 CAD
CA76683-154 CA76683-156
sparQ mRNA-Seq Kit
Vectors, Plasmids and Libraries
Ultra-sensitive mRNA library prep workflow with efficient poly(A) capture.
  • Highly sensitive mRNA detection with input as low as 5 ng of total RNA
  • Reproducible transcript profiling
  • High quality directional RNA library prep with streamlined workflow of 4.75 hours
  • Efficient isolation of mRNA allowing detection of most coding genes
sparQ mRNA-Seq Kit enables efficient preparation of stranded mRNA-seq libraries for Illumina® NGS platforms. The streamlined workflow reduces hands-on time and allows sample to sequencing in a single day. High-quality, reliable mRNA-seq libraries are delivered with consistency and reduced bias from as little as 5 ng and up to 1 μg of total RNA.
Packaging: Store Box 1 in a constant temperature refrigerator at 2°C to 8°C upon receipt. Store Box 2 in a constant temperature freezer at -25°C to -15°C upon receipt.