Cryogenic Storage Labels
Supplier: Computype
1061484 1061485 1061487 1061488 1061490 1061491
76635-404KT 1032.67 CAD
76635-404 76635-406 76635-408 76635-410 76635-412 76635-414
Cryogenic Storage Labels
Labels
Wrap-around labels for glass or plastic tubes and vials exposed to temperatures as low as −196 °C.
  • TS1113 polypropylene
  • TR618Z ribbon: 52×153 mm
  • Self-adherese and completely wraps around tubes ranging in size from 0.5 to 50 ml
  • Withstands both direct and vapor liquid nitrogen exposure and an autoclave
  • Resistant to solvents such as DMSO, methanol, isopropyl alcohol and more
Designed to protect both print and adhesion, these labels wrap completely around tubes of ranging sizes and self-adhere. The self-laminating design protects sample integrity throughMore Product Information