Logic® Vue Canopy Connection Kits
Supplier: Labconco
3347811 3347813
76645-800EA 1919.7 CAD
76645-800 76645-804
Logic® Vue Canopy Connection Kits
Fume Hoods and Enclosures Biological Safety Cabinets
Logic® Vue canopy connection kit for thimble ducting of 4 or 6' Logic Vue class II enclosures.
  • Connects Logic Vue to laboratory exhaust system
  • Maintains safe, negative pressure exhast flow
  • Built-in alarm system - Sends audible and visual alarm from Logic Vue
  • Built-in exhaust sensor - Detects remote exhaust failure in <1 second
  • Includes 12" ID duct collar
For thimble exhausting the 4 or 6' Logic Vue. Canopy maintains a negative presssure connection to house exhaust. Detects insufficent exaust in less than one second and signals MyLogic™ OS to display and sound alarm. Exhaust connection and remote exhasust required. Exhaust damper recommended.