Dispenser Dropping Bottles
Supplier: United Scientific Supplies
UNDDB015 UNDDB030 UNDDB060 UNDDB125 UNDDB250
470349-746PK 82.5 CAD
470349-746 470349-766 470349-716 470349-694 470349-768
Dispenser Dropping Bottles
Bottles Dropping Bottles
Precision dispensing of solutions.
  • Dispenses one drop at a time
  • Spout lid is connected to the cap
  • LDPE with leakproof PP cap
  • BPA-free
This LDPE dispensing bottle can be easily operated with one hand. The bottle features a polypropylene screw cap with a tethered spout lid that snaps securely in place.